siRNA Details
Gene Symbol: RGD1307315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008377.1 | NP_001008378.1 | 3 | 759 | ||
GenBank mRNA | BC086564.1 | - | 3 | 759 | 7305233 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1307315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008377.1 | NP_001008378.1 | 3 | 759 | ||
GenBank mRNA | BC086564.1 | - | 3 | 759 | 7305233 |