siRNA Details
| Gene Symbol: Nabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034939.1 | NP_001030111.2 | 3 | 641 | ||
| NM_001244819.1 | NP_001231748.1 | 4 | 364 | |||
| XM_006240769.2 | XP_006240831.1 | 4 | 605 | |||
| XM_006240770.2 | XP_006240832.1 | 4 | 460 | |||
| XM_006240771.2 | XP_006240833.1 | 4 | 455 | |||
| XM_008765029.1 | XP_008763251.1 | 4 | 620 | |||
| GenBank mRNA | BC104710.1 | - | 3 | 641 | 7319804 | |
| DV214819.1 | - | 4 | 364 | |||
| FQ219805.1 | - | 4 | 352 | |||

Nabp2
Genomic Map