siRNA Details
| Gene Symbol: Rnf41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012195.1 | NP_001012195.1 | 6 | 801 | ||
| XM_006240772.2 | XP_006240834.1 | 8 | 1232 | |||
| XM_006240773.2 | XP_006240835.1 | 8 | 1294 | |||
| XM_006240774.2 | XP_006240836.1 | 7 | 1140 | |||
| XM_006240775.2 | XP_006240837.1 | 8 | 1150 | |||
| XM_006240776.2 | XP_006240838.1 | 7 | 1058 | |||
| XM_008765030.1 | XP_008763252.1 | 7 | 2151 | |||
| GenBank mRNA | BC083614.1 | - | 6 | 801 | 7133774 | |

Rnf41
Genomic Map