siRNA Details
| Gene Symbol: Pmel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240779.2 | XP_006240841.1 | 6 | 1566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pmel| Gene Symbol: Pmel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240779.2 | XP_006240841.1 | 6 | 1566 | ||