siRNA Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033070.1 | NP_001028242.1 | 2 | 251 | ||
| XM_006240781.2 | XP_006240843.1 | 2 | 280 | |||
| XM_008765031.1 | XP_008763253.1 | 2 | 275 | |||
| GenBank mRNA | BC100070.1 | - | 2 | 251 | 7385206 | |

Sarnp
Genomic Map