siRNA Details
| Gene Symbol: Atcay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040190.1 | NP_001035280.1 | 8, 9 | 1350 | ||
| GenBank mRNA | AY611623.1 | - | 8, 9 | 1350 | ||
| BC128695.1 | - | 8, 9 | 1202 | 8366437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atcay| Gene Symbol: Atcay | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040190.1 | NP_001035280.1 | 8, 9 | 1350 | ||
| GenBank mRNA | AY611623.1 | - | 8, 9 | 1350 | ||
| BC128695.1 | - | 8, 9 | 1202 | 8366437 | ||