siRNA Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108735.1 | NP_001102205.1 | 2 | 428 | ||
| XM_006240965.1 | XP_006241027.1 | 2 | 422 | |||
| XM_006240966.1 | XP_006241028.1 | 2 | 321 | |||
| XM_008765119.1 | XP_008763341.1 | 3 | 216 | |||
| XM_008765120.1 | XP_008763342.1 | 2 | 156 | |||
| XM_008765121.1 | XP_008763343.1 | 3 | 224 | |||
| XM_008765122.1 | XP_008763344.1 | 2 | 336 | |||
| GenBank mRNA | BC167041.1 | - | 2 | 195 | ||

Mbd3
Genomic Map