siRNA Details
| Gene Symbol: R3hdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173974.1 | NP_001167445.1 | 2, 3 | 267 | ||
| GenBank mRNA | BC161971.1 | - | 2, 3 | 267 | 9028992 | |
| FQ213685.1 | - | 2, 3 | 275 | |||
| FQ228736.1 | - | 2, 3 | 243 | |||

R3hdm4
Genomic Map