siRNA Details
| Gene Symbol: Zfp870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163062.1 | NP_001156534.1 | 4 | 1375 | ||
| XM_006241082.2 | XP_006241144.1 | 4 | 1462 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp870| Gene Symbol: Zfp870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163062.1 | NP_001156534.1 | 4 | 1375 | ||
| XM_006241082.2 | XP_006241144.1 | 4 | 1462 | |||