siRNA Details
| Gene Symbol: Angptl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199115.2 | NP_954546.1 | 7 | 1297 | ||
| GenBank mRNA | AY393999.1 | - | 7 | 1157 | ||
| BC078944.1 | - | 7 | 1297 | 7109431 | ||
| FQ210814.1 | - | 2 | 314 | |||
| FQ218836.1 | - | 4 | 567 | |||

Angptl4
Genomic Map