siRNA Details
| Gene Symbol: Angptl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199115.2 | NP_954546.1 | 2 | 540 | ||
| GenBank mRNA | AY393999.1 | - | 2 | 400 | ||
| BC078944.1 | - | 2 | 540 | 7109431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Angptl4| Gene Symbol: Angptl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199115.2 | NP_954546.1 | 2 | 540 | ||
| GenBank mRNA | AY393999.1 | - | 2 | 400 | ||
| BC078944.1 | - | 2 | 540 | 7109431 | ||