siRNA Details
Gene Symbol: Tmem263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108739.1 | NP_001102209.1 | 3 | 361 | ||
XM_006241164.2 | XP_006241226.1 | 3 | 394 | |||
XM_008765244.1 | XP_008763466.1 | 3 | 984 | |||
GenBank mRNA | BC166979.1 | - | 3 | 264 | 7315264 | |
FQ218051.1 | - | 1 | 171 |

More Information
Other Species Targeted

