siRNA Details
| Gene Symbol: Appl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108741.1 | NP_001102211.1 | 21 | 1985 | ||
| GenBank mRNA | BC168167.1 | - | 21 | 1900 | 9021772 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Appl2| Gene Symbol: Appl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108741.1 | NP_001102211.1 | 21 | 1985 | ||
| GenBank mRNA | BC168167.1 | - | 21 | 1900 | 9021772 | |