siRNA Details
| Gene Symbol: Myrfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765434.1 | XP_008763656.1 | 28 | 3361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myrfl| Gene Symbol: Myrfl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765434.1 | XP_008763656.1 | 28 | 3361 | ||