siRNA Details
| Gene Symbol: RGD1306474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108746.1 | NP_001102216.1 | 2, 3 | 379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1306474| Gene Symbol: RGD1306474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108746.1 | NP_001102216.1 | 2, 3 | 379 | ||