siRNA Details
| Gene Symbol: Tmbim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199116.2 | NP_954547.1 | 3 | 352 | ||
| GenBank mRNA | BC060596.1 | - | 3 | 353 | 6889044 | |
| FQ217672.1 | - | 3 | 295 | |||
| FQ228009.1 | - | 3 | 349 | |||
| FQ230396.1 | - | 3 | 317 | |||
| FQ233689.1 | - | 3 | 346 | |||

Tmbim4
Genomic Map