siRNA Details
| Gene Symbol: Tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008378.1 | NP_001008379.1 | 1, 2 | 149 | ||
| GenBank mRNA | BC086452.1 | - | 1, 2 | 149 | 7307024 | |
| FQ214182.1 | - | 1, 2 | 119 | |||
| FQ218380.1 | - | 1, 2 | 119 | |||
| FQ219469.1 | - | 1, 2 | 142 | |||
| FQ219661.1 | - | 1, 2 | 147 | |||

Tspan31
Genomic Map