siRNA Details
| Gene Symbol: Arhgap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012198.1 | NP_001012198.1 | 14 | 1213 | ||
| NM_001080789.1 | NP_001074258.1 | 15 | 1845 | |||
| XM_006241464.2 | XP_006241526.1 | 15 | 1932 | |||
| XM_006241465.2 | XP_006241527.1 | 15 | 1923 | |||
| XM_006241466.2 | XP_006241528.1 | 15 | 1966 | |||
| XM_006241468.2 | XP_006241530.1 | 14 | 1270 | |||
| GenBank mRNA | BC079334.1 | - | 14 | 1213 | 7097179 | |
| BC107938.1 | - | 15 | 1845 | 7378387 | ||

Arhgap9
Genomic Map