siRNA Details
| Gene Symbol: Stac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130558.1 | NP_001124030.1 | 3 | 382 | ||
| XM_006241486.2 | XP_006241548.1 | 3 | 895 | |||
| XM_006241488.2 | XP_006241550.1 | 2 | 827 | |||
| XM_008765415.1 | XP_008763637.1 | 3 | 317 | |||
| GenBank mRNA | FQ216646.1 | - | 2 | 206 | ||

Stac3
Genomic Map