siRNA Details
Gene Symbol: LOC362901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008877.2 | NP_001008877.1 | 2 | 139 | ||
GenBank mRNA | AY310150.1 | - | 179 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC362901 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008877.2 | NP_001008877.1 | 2 | 139 | ||
GenBank mRNA | AY310150.1 | - | 179 |