siRNA Details
| Gene Symbol: Wdyhv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025024.1 | NP_001020195.1 | 2 | 256 | ||
| GenBank mRNA | BC091306.1 | - | 2 | 256 | 7311328 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdyhv1| Gene Symbol: Wdyhv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025024.1 | NP_001020195.1 | 2 | 256 | ||
| GenBank mRNA | BC091306.1 | - | 2 | 256 | 7311328 | |