siRNA Details
| Gene Symbol: Anxa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134910.1 | NP_001128382.1 | 9 | 738 | ||
| XM_008765512.1 | XP_008763734.1 | 9 | 1070 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anxa13| Gene Symbol: Anxa13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134910.1 | NP_001128382.1 | 9 | 738 | ||
| XM_008765512.1 | XP_008763734.1 | 9 | 1070 | |||