siRNA Details
| Gene Symbol: Mtss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130563.1 | NP_001124035.1 | 15, 16 | 1829 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtss1| Gene Symbol: Mtss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130563.1 | NP_001124035.1 | 15, 16 | 1829 | ||