siRNA Details
| Gene Symbol: St3gal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013219.1 | NP_001013237.1 | 2 | 362 | ||
| XM_006241709.2 | XP_006241771.1 | 5 | 1334 | |||
| XM_006241710.2 | XP_006241772.1 | 5 | 2252 | |||
| XM_008765513.1 | XP_008763735.1 | 4 | 1279 | |||
| GenBank mRNA | AJ748840.1 | - | 2 | 362 | ||

St3gal1
Genomic Map