siRNA Details
| Gene Symbol: Zfat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134957.1 | NP_001128429.1 | 3 | 410 | ||
| XM_006241712.2 | XP_006241774.1 | 3 | 2545 | |||
| XM_006241713.2 | XP_006241775.1 | 3 | 2545 | |||
| XM_006241714.2 | XP_006241776.1 | 3 | 2545 | |||
| XM_008765515.1 | XP_008763737.1 | 3 | 523 | |||

Zfat
Genomic Map