siRNA Details
Gene Symbol: Zfp647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_343279.7 | XP_343280.3 | 5 | 1243 |
Gene Symbol: LOC102555083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241920.2 | XP_006241982.1 | 1243 |
