siRNA Details
| Gene Symbol: Nol12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012747.2 | NP_001012765.1 | 1 | 80 | ||
| GenBank mRNA | AY917132.1 | - | 1 | 58 | ||
| BC103643.1 | - | 1 | 80 | 7301307 | ||
| FQ227943.1 | - | 1 | 128 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nol12| Gene Symbol: Nol12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012747.2 | NP_001012765.1 | 1 | 80 | ||
| GenBank mRNA | AY917132.1 | - | 1 | 58 | ||
| BC103643.1 | - | 1 | 80 | 7301307 | ||
| FQ227943.1 | - | 1 | 128 | |||