siRNA Details
| Gene Symbol: Triobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013220.2 | NP_001013238.2 | 18 | 5453 | ||
| XM_006242010.2 | XP_006242072.1 | 9 | 1292 | |||
| XM_008765758.1 | XP_008763980.1 | 9 | 1556 | |||
| XM_008765759.1 | XP_008763981.1 | 9 | 1556 | |||
| GenBank mRNA | BC169041.1 | - | 9 | 1232 | ||
| EF203432.1 | - | 8 | 1142 | |||
| EF203433.1 | - | 8 | 1142 | |||

Triobp
Genomic Map