siRNA Details
| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025285.1 | NP_001020456.1 | 3 | 474 | ||
| XM_006242012.2 | XP_006242074.1 | 3 | 547 | |||
| GenBank mRNA | BC093616.1 | - | 3 | 474 | 7311744 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd54| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025285.1 | NP_001020456.1 | 3 | 474 | ||
| XM_006242012.2 | XP_006242074.1 | 3 | 547 | |||
| GenBank mRNA | BC093616.1 | - | 3 | 474 | 7311744 | |