siRNA Details
| Gene Symbol: Tmem184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173370.1 | NP_001166841.1 | 8 | 1129 | ||
| XM_006242013.2 | XP_006242075.1 | 8 | 1136 | |||
| XM_006242014.2 | XP_006242076.1 | 8 | 1722 | |||
| GenBank mRNA | BC099785.1 | - | 8 | 1129 | ||

Tmem184b
Genomic Map