siRNA Details
| Gene Symbol: Dmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130567.1 | NP_001124039.1 | 10 | 1108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmc1| Gene Symbol: Dmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130567.1 | NP_001124039.1 | 10 | 1108 | ||