siRNA Details
| Gene Symbol: Nfam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242140.2 | XP_006242202.1 | 4 | 719 | ||
| XM_006242142.2 | XP_006242204.1 | 4 | 632 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nfam1| Gene Symbol: Nfam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242140.2 | XP_006242202.1 | 4 | 719 | ||
| XM_006242142.2 | XP_006242204.1 | 4 | 632 | |||