siRNA Details
| Gene Symbol: Creld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037208.1 | NP_001032285.1 | 7 | 784 | ||
| XM_006242216.2 | XP_006242278.1 | 6 | 639 | |||
| GenBank mRNA | BC098722.1 | - | 7 | 784 | 7371287 | |
| FQ229732.1 | - | 3 | 352 | |||

Creld2
Genomic Map