siRNA Details
| Gene Symbol: Creld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037208.1 | NP_001032285.1 | 9 | 923 | ||
| XM_006242216.2 | XP_006242278.1 | 8 | 778 | |||
| GenBank mRNA | BC098722.1 | - | 9 | 923 | 7371287 | |
| FQ229732.1 | - | 5 | 491 | |||

Creld2
Genomic Map