siRNA Details
| Gene Symbol: Hdac10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035000.1 | NP_001030172.1 | 6 | 768 | ||
| XM_006242218.1 | XP_006242280.1 | 6 | 1677 | |||
| XM_006242219.1 | XP_006242281.1 | 6 | 1677 | |||
| XM_006242220.1 | XP_006242282.1 | 6 | 1677 | |||
| GenBank mRNA | BC092573.1 | - | 6 | 768 | 7323327 | |

Hdac10
Genomic Map