siRNA Details
| Gene Symbol: Cpne8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108750.1 | NP_001102220.1 | 12, 13 | 1013 | ||
| GenBank mRNA | AM747284.1 | - | 12, 13 | 884 | ||
| BC168840.1 | - | 12, 13 | 936 | 7125016 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpne8| Gene Symbol: Cpne8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108750.1 | NP_001102220.1 | 12, 13 | 1013 | ||
| GenBank mRNA | AM747284.1 | - | 12, 13 | 884 | ||
| BC168840.1 | - | 12, 13 | 936 | 7125016 | ||