siRNA Details
| Gene Symbol: Ccdc65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014203.1 | NP_001014225.1 | 7 | 1198 | ||
| GenBank mRNA | BC083683.1 | - | 7 | 1198 | 7135919 | |
| FQ212708.1 | - | 5 | 960 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc65| Gene Symbol: Ccdc65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014203.1 | NP_001014225.1 | 7 | 1198 | ||
| GenBank mRNA | BC083683.1 | - | 7 | 1198 | 7135919 | |
| FQ212708.1 | - | 5 | 960 | |||