siRNA Details
| Gene Symbol: Dnajc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014204.1 | NP_001014226.1 | 2 | 561 | ||
| XM_006257363.2 | XP_006257425.1 | 2 | 823 | |||
| GenBank mRNA | BC086949.1 | - | 2 | 561 | 7325189 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajc22| Gene Symbol: Dnajc22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014204.1 | NP_001014226.1 | 2 | 561 | ||
| XM_006257363.2 | XP_006257425.1 | 2 | 823 | |||
| GenBank mRNA | BC086949.1 | - | 2 | 561 | 7325189 | |