siRNA Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108752.1 | NP_001102222.1 | 4, 5 | 537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smarcd1| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108752.1 | NP_001102222.1 | 4, 5 | 537 | ||