siRNA Details
| Gene Symbol: Tarbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034941.1 | NP_001030113.1 | 2 | 249 | ||
| XM_006242428.2 | XP_006242490.1 | 2 | 329 | |||
| XM_006242429.2 | XP_006242491.1 | 2 | 262 | |||
| GenBank mRNA | BC104690.1 | - | 2 | 249 | 7107060 | |
| FQ228813.1 | - | 2 | 243 | |||

Tarbp2
Genomic Map