siRNA Details
| Gene Symbol: Mrpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108754.1 | NP_001102224.1 | 4, 5 | 526 | ||
| GenBank mRNA | FQ215152.1 | - | 4, 5 | 392 | ||
| FQ226445.1 | - | 4, 5 | 392 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl4| Gene Symbol: Mrpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108754.1 | NP_001102224.1 | 4, 5 | 526 | ||
| GenBank mRNA | FQ215152.1 | - | 4, 5 | 392 | ||
| FQ226445.1 | - | 4, 5 | 392 | |||