siRNA Details
| Gene Symbol: Slc44a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242649.2 | XP_006242711.1 | 22 | 2896 | ||
| XM_006242650.2 | XP_006242712.1 | 22 | 2166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc44a2| Gene Symbol: Slc44a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242649.2 | XP_006242711.1 | 22 | 2896 | ||
| XM_006242650.2 | XP_006242712.1 | 22 | 2166 | |||