siRNA Details
| Gene Symbol: Yipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014208.1 | NP_001014230.1 | 4 | 347 | ||
| GenBank mRNA | BC083587.1 | - | 4 | 347 | 7131986 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yipf2| Gene Symbol: Yipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014208.1 | NP_001014230.1 | 4 | 347 | ||
| GenBank mRNA | BC083587.1 | - | 4 | 347 | 7131986 | |