siRNA Details
Gene Symbol: Yipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014208.1 | NP_001014230.1 | 5 | 454 | ||
GenBank mRNA | BC083587.1 | - | 5 | 454 | 7131986 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Yipf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014208.1 | NP_001014230.1 | 5 | 454 | ||
GenBank mRNA | BC083587.1 | - | 5 | 454 | 7131986 |