siRNA Details
Gene Symbol: Spc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750448.3 | XP_003750496.3 | 3, 4 | 695 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750448.3 | XP_003750496.3 | 3, 4 | 695 |