siRNA Details
| Gene Symbol: Rp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108756.1 | NP_001102226.1 | 3 | 358 | ||
| XM_008765960.1 | XP_008764182.1 | 3 | 452 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rp9| Gene Symbol: Rp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108756.1 | NP_001102226.1 | 3 | 358 | ||
| XM_008765960.1 | XP_008764182.1 | 3 | 452 | |||