siRNA Details
| Gene Symbol: Pate4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108757.1 | NP_001102227.1 | 2 | 109 | ||
| XM_008766061.1 | XP_008764283.1 | 1 | 243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pate4| Gene Symbol: Pate4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108757.1 | NP_001102227.1 | 2 | 109 | ||
| XM_008766061.1 | XP_008764283.1 | 1 | 243 | |||