siRNA Details
| Gene Symbol: Cadm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012201.1 | NP_001012201.1 | 2 | 330 | ||
| XM_006242944.2 | XP_006243006.1 | 2 | 329 | |||
| XM_006242946.2 | XP_006243008.1 | 2 | 329 | |||
| XM_006242947.2 | XP_006243009.1 | 2 | 329 | |||
| XM_006242948.2 | XP_006243010.1 | 2 | 329 | |||
| GenBank mRNA | AB114443.1 | - | 2 | 219 | ||
| AB257091.1 | - | 2 | 167 | |||
| BC078966.1 | - | 2 | 330 | 7111523 | ||

Cadm1
Genomic Map