siRNA Details
| Gene Symbol: Zw10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024801.1 | NP_001019972.1 | 3 | 354 | ||
| GenBank mRNA | BC097452.1 | - | 3 | 354 | 7456845 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zw10| Gene Symbol: Zw10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024801.1 | NP_001019972.1 | 3 | 354 | ||
| GenBank mRNA | BC097452.1 | - | 3 | 354 | 7456845 | |