siRNA Details
| Gene Symbol: Ube2q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766429.1 | XP_008764651.1 | 1, 2 | 513 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2q2| Gene Symbol: Ube2q2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766429.1 | XP_008764651.1 | 1, 2 | 513 | ||